Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYST1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055251F
Description
MYST1 Polyclonal antibody specifically detects MYST1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MYST1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human males absent on the first (MYST histone acetyltransferase 1) using the numbering given in entry BAB13924.1 (GeneID 84148). | |
0.1 mL | |
Chromatin Research, DNA Repair | |
84148 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction