Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578596 Supplier Invitrogen™ Supplier No. PA578596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, 293T NE, A431, HeLa, HepG2 Predicted Reactivity: Mouse (100%), Rat (100%), Zebrafish (84%), Pig (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYST1
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene KAT8
Gene Accession No. Q9H7Z6
Gene Alias 2010203C02Rik; 5830450F21Rik; D7Ertd629e; h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase Myst1; hMOF; K (lysine) acetyltransferase 8; K(lysine) acetyltransferase 8; Kat8; Lysine acetyltransferase 8; MOF; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST histone acetyltransferase 1; MYST protein 1; MYST1; MYST-1; ortholog of Drosophila males absent on the first (MOF); PP7073; probable histone acetyltransferase MYST1; ZC2HC8
Gene Symbols KAT8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MYST1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84148
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.