Learn More
Invitrogen™ MYST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596098
Description
Immunogen sequence: MAAQGAAAAV AAGTSGVAGE GEPGPGENAA AEGTAPSPGR VSPPTPARGE PEVTVEIGET YLCRRPDSTW HSAEVIQSRV NDQEGREEFY VHYVGFNRRL DEWVDKNRLA LTKTVKDAVQ KNSEKYLSEL AEQPERKITR NQKRKHDEIN HVQKTYAEMD PTTAALEKEH EAITKVKYVD KIHIGNYEID AWYFSPFPED; Positive Samples: OVCAR3, NIH/3T3; Cellular Location: Chromosome, Nucleus.
The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.
Specifications
MYST1 | |
Polyclonal | |
Unconjugated | |
KAT8 | |
2010203C02Rik; 5830450F21Rik; D7Ertd629e; h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase Myst1; hMOF; K (lysine) acetyltransferase 8; K(lysine) acetyltransferase 8; Kat8; Lysine acetyltransferase 8; MOF; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST histone acetyltransferase 1; MYST protein 1; MYST1; MYST-1; ortholog of Drosophila males absent on the first (MOF); PP7073; probable histone acetyltransferase MYST1; ZC2HC8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
310194, 67773, 84148 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.76 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q5XI06, Q9D1P2, Q9H7Z6 | |
KAT8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human KAT8/MYST1/MOF (NP_8920032). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.