Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYST1 Recombinant Rabbit Monoclonal Antibody (6U5Z6)
GREENER_CHOICE

Catalog No. PIMA538064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538064 Supplier Invitrogen™ Supplier No. MA538064
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HepG2, MCF7, NIH/3T3 Immunogen sequence: EKPLSDLGKL SYRSYWSWVL LEILRDFRGT LSIKDLSQMT SITQNDIIST LQSLNMVKYW KGQHVICVTP KLVEEHLKSA QYKKPPITVD SVCLKWAPPK HKQVKLSKK.

The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYST1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6U5Z6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene KAT8
Gene Accession No. Q9D1P2, Q9H7Z6
Gene Alias 2010203C02Rik; 5830450F21Rik; D7Ertd629e; h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase Myst1; hMOF; K (lysine) acetyltransferase 8; K(lysine) acetyltransferase 8; Kat8; Lysine acetyltransferase 8; MOF; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST histone acetyltransferase 1; MYST protein 1; MYST1; MYST-1; ortholog of Drosophila males absent on the first (MOF); PP7073; probable histone acetyltransferase MYST1; ZC2HC8
Gene Symbols KAT8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-458 of human KAT8/MYST1/MOF (Q9H7Z6).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67773, 84148
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.