Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myt1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299008MFV610
Description
Myt1 Polyclonal antibody specifically detects Myt1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Myt1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
C20orf36, MTF1proteolipid protein binding protein, myelin transcription factor 1, Myelin transcription factor I, MyT1, MyTI, MYTIKIAA0835, PLPB1KIAA1050, Proteolipid protein-binding protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Myt1. | |
0.1 mL | |
Neuroscience, Protein Kinase | |
4661 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction