Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576519
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported. The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system.
Specifications
MYT1 | |
Polyclonal | |
Unconjugated | |
MYT1 | |
C20orf36; KIAA0835; KIAA1050; MTF1; myelin transcription factor 1; Myelin transcription factor I; MYT1; MyTI; Neural zinc finger factor 2; neural zinc finger transcription factor 2; Nzf2; NZF-2; Nzf2a; Nzf2b; Nztf2; PLPB1; proteolipid protein binding protein; proteolipid protein-binding protein; ZC2H2C1; ZC2HC4A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17932, 362291, 4661 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q01538, Q8CFC2 | |
MYT1 | |
Recombinant fusion protein of human MYT1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction