Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588052 Supplier Invitrogen™ Supplier No. PA588052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPQDLPSKSV DIEVDENGTL DLSMHKHRKR ENAFPSSSSC SSSPGVKSPD ASQRHSSTSA PSSSMTSPQS SQASRQDEWD RPLDYTKPSR LREEEPEESE PAAHSFASSE ADDQEVSEEN FEERKYPGEV TLTNFKLKFL SKDIKKELLT CPTPGCDGSG HITGNYASHR S; Positive Samples: U-87MG, U-251MG, Mouse liver, Mouse kidney, Rat kidney, Rat liver; Cellular Location: Nucleus.

The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported. The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MYT1
Gene Accession No. Q01538, Q8CFC2
Gene Alias C20orf36; KIAA0835; KIAA1050; MTF1; myelin transcription factor 1; Myelin transcription factor I; MYT1; MyTI; Neural zinc finger factor 2; neural zinc finger transcription factor 2; Nzf2; NZF-2; Nzf2a; Nzf2b; Nztf2; PLPB1; proteolipid protein binding protein; proteolipid protein-binding protein; ZC2H2C1; ZC2HC4A
Gene Symbols MYT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 650-820 of human MYT1 (NP_0045261).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17932, 362291, 4661
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.