Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYT1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534468
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534468 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534468 Supplier Invitrogen™ Supplier No. PA534468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse brain tissue lysate. PA5-34468 can be used with blocking peptide PEP-1510.

MYT1L, myelin transcription factor 1 family, to which the myelin transcription factor 1-like protein (MYT1L) belongs, is expressed primarily in the developing central nervous system and recruits histone deacetylase (HDAC) to regulate neural transcription. Both MYT1L and the related protein MYT1 interact with SIN3B, a protein that mediates transcriptional repression by binding to HDACs, suggesting that the MYT1 family favor the silencing of genes during neural development. Recent studies suggest that polymorphisms of MYT1L may be associated with schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYT1L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MYT1L
Gene Accession No. P70475, P97500, Q9UL68
Gene Alias 2900046C06Rik; 2900093J19Rik; C630034G21Rik; Kiaa1106; mKIAA1106; MRD39; myelin transcription factor 1 like; myelin transcription factor 1-like; myelin transcription factor 1-like protein; Myt1l; MyT1-L; Neural zinc finger factor 1; neural zinc finger protein NZF-1; neural zinc finger transcription factor 1; Nzf1; NZF-1; Nztf1; Pmng1; Png1; Png-1; postmeiotic neural gene 1; postmitotic neural gene 1 protein; ZC2H2C2; ZC2HC4B; zinc finger protein Png-1
Gene Symbols MYT1L
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human MYT1L.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116668, 17933, 23040
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.