Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYZAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558946 Supplier Invitrogen™ Supplier No. PA558946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVPPESPVPE QCEKKIERKE QLLDLSNGEP TRKLPQGVVY GVVRRSDQNQ QKEMVVYGWS TSQLKEEMNY IKDVRATLEK VRKRMYGDYD EM Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

MYOZAP, also known as GRINL1A, is a 54 kDa highly conserved cardiac protein. It is strongly expressed in the heart and lung and is a novel component of intercalated disc. MYOZAP interacts with myosin phosphatase-RhoA interacting protein (MRIP) and acts as an activator of Rho-dependent SRF signaling. Knockdown study in zebrafish results in cardiomyopathy with severe dysfunction. The MYOZAP gene is part of a complex transcript unit that includes the gene for glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A (GRINL1A). Transcription of this gene occurs at an upstream promoter, with two different groups of alternatively spliced variants: Gup for GRINL1A upstream transcripts and Gcom for GRINL1A combined transcripts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYZAP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYZAP
Gene Accession No. P0CAP1
Gene Alias AA407270; AV006038; gcom; GCOM1; Gcom2; Gm640; GRINL1A; GRINL1A combined protein; GRINL1A complex locus 1; GRINL1A complex locus protein 1; GRINL1A complex locus upstream; GRINL1A upstream protein; Grinl1a7; Gup; myocardial intercalated disc protein; myocardial zonula adherens protein; myocardium-enriched ZO1-associated protein; myocardium-enriched zonula adherens protein; myocardium-enriched zonula occludens-1-associated protein; myozap; Myzap; MYZAP-POLR2M
Gene Symbols MYZAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYZAP. Recombinant protein control fragment (Product #RP-98067).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100820829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.