Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MZB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584122 Supplier Invitrogen™ Supplier No. PA584122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TATAPQLDDE EMYSAHMPAH LRCDACRAVA YQMWQNLAKA ETKLHTSNSG GRRELSELVY TDVLDRS Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 72%.

MZB1 associates with immunoglobulin M (IgM) heavy and light chains and promotes IgM assembly and secretion. May exert its effect by acting as a molecular chaperone or as an oxidoreductase as it displays a low level of oxidoreductase activity. Isoform 2 may be involved in regulation of apoptosis. Helps to diversify peripheral B-cell functions by regulating Ca(2+) stores, antibody secretion and integrin activation.; Acts as a hormone-regulated adipokine/proinflammatory cytokine that is implicated in causing chronic inflammation, affecting cellular expansion and blunting insulin response in adipocytes. May have a role in the onset of insulin resistance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MZB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MZB1
Gene Accession No. Q8WU39
Gene Alias caspase-2 binding protein; HSPC190; marginal zone B and B1 cell specific protein; marginal zone B and B1 cell-specific protein; marginal zone B- and B1-cell-specific protein; MEDA7; MEDA-7; Mesenteric estrogen-dependent adipose 7; mesenteric oestrogen-dependent adipose gene- 7; MGC29506; MZB1; PACAP; pERp1; plasma cell-induced ER protein 1; plasma cell-induced resident endoplasmic reticulum protein; Plasma cell-induced resident ER protein; proapoptotic caspase adapter protein; proapoptotic caspase adaptor protein
Gene Symbols MZB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MZB1. Recombinant protein control fragment (Product #RP-104951).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.