Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MZB1 Recombinant Rabbit Monoclonal Antibody (022)
GREENER_CHOICE

Catalog No. PIMA529430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529430 Supplier Invitrogen™ Supplier No. MA529430
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human PERP1/MZB1.

MZB1 associates with immunoglobulin M (IgM) heavy and light chains and promotes IgM assembly and secretion. May exert its effect by acting as a molecular chaperone or as an oxidoreductase as it displays a low level of oxidoreductase activity. Isoform 2 may be involved in regulation of apoptosis. Helps to diversify peripheral B-cell functions by regulating Ca(2+) stores, antibody secretion and integrin activation.; Acts as a hormone-regulated adipokine/proinflammatory cytokine that is implicated in causing chronic inflammation, affecting cellular expansion and blunting insulin response in adipocytes. May have a role in the onset of insulin resistance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MZB1
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 022
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MZB1
Gene Accession No. Q8WU39
Gene Alias caspase-2 binding protein; HSPC190; marginal zone B and B1 cell specific protein; marginal zone B and B1 cell-specific protein; marginal zone B- and B1-cell-specific protein; MEDA7; MEDA-7; Mesenteric estrogen-dependent adipose 7; mesenteric oestrogen-dependent adipose gene- 7; MGC29506; MZB1; PACAP; pERp1; plasma cell-induced ER protein 1; plasma cell-induced resident endoplasmic reticulum protein; Plasma cell-induced resident ER protein; proapoptotic caspase adapter protein; proapoptotic caspase adaptor protein
Gene Symbols MZB1
Host Species Rabbit
Immunogen Recombinant Human PERP1/MZB1 Protein (Met 1- Thr 185).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.