Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MZF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551018
Description
MZF1 Polyclonal Antibody detects endogenous levels of MZF1 protein.
Binds to target promoter DNA and functions as transcription regulator. Regulates transcription from the PADI1 and CDH2 promoter. May be one regulator of transcriptional events during hemopoietic development. [UniProt].
Specifications
MZF1 | |
Polyclonal | |
Unconjugated | |
MZF1 | |
myeloid zinc finger 1; Myeloid zinc finger protein 1; Myeloid zinc finger protein 2; myeloid-specific retinoic acid-responsive zinc finger protein; MZF; MZF1; MZF-1; MZF1B; Mzf2; Mzf-2; Zfp121; Zfp98; Zinc finger and SCAN domain-containing protein 6; zinc finger protein 121; Zinc finger protein 42; zinc finger protein 42 (myeloid-specific retinoic acid-responsive); zinc finger protein 98; Znf42; ZSCAN6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7593 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P28698 | |
MZF1 | |
Synthesized peptide derived from the N-terminal region of human MZF-1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction