Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MZF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551630 Supplier Invitrogen™ Supplier No. PA551630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLTQHLRVHT GEKPFACPEC GQRFSQRLKL TRHQRTHTGE KPYHCGECGL GFTQVSRLTE HQRIHTGERP FACPECGQSF RQHANLTQHR RIHTGERPYA CPECGKAFRQ RPTLTQHLRT HRREKPFACQ DCGRRFHQST K Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Binds to target promoter DNA and functions as transcription regulator. Regulates transcription from the PADI1 and CDH2 promoter. May be one regulator of transcriptional events during hemopoietic development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MZF1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MZF1
Gene Accession No. P28698
Gene Alias myeloid zinc finger 1; Myeloid zinc finger protein 1; Myeloid zinc finger protein 2; myeloid-specific retinoic acid-responsive zinc finger protein; MZF; MZF1; MZF-1; MZF1B; Mzf2; Mzf-2; Zfp121; Zfp98; Zinc finger and SCAN domain-containing protein 6; zinc finger protein 121; Zinc finger protein 42; zinc finger protein 42 (myeloid-specific retinoic acid-responsive); zinc finger protein 98; Znf42; ZSCAN6
Gene Symbols MZF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MZF1. Recombinant protein control fragment (Product #RP-100724).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7593
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.