Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MZF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106478 Supplier Invitrogen™ Supplier No. PA5106478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MZF-1.

Binds to target promoter DNA and functions as transcription regulator. Regulates transcription from the PADI1 and CDH2 promoter. May be one regulator of transcriptional events during hemopoietic development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MZF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MZF1
Gene Accession No. P28698
Gene Alias myeloid zinc finger 1; Myeloid zinc finger protein 1; Myeloid zinc finger protein 2; myeloid-specific retinoic acid-responsive zinc finger protein; MZF; MZF1; MZF-1; MZF1B; Mzf2; Mzf-2; Zfp121; Zfp98; Zinc finger and SCAN domain-containing protein 6; zinc finger protein 121; Zinc finger protein 42; zinc finger protein 42 (myeloid-specific retinoic acid-responsive); zinc finger protein 98; Znf42; ZSCAN6
Gene Symbols MZF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MZF1(Accession P28698), corresponding to amino acid residues M1-P40.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109889, 361508, 7593
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.