Learn More
Invitrogen™ MZF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120086
Description
Positive test controls include: HeLa, A-431, U-251MG, 293T, SGC-7901, Mouse brain, Mouse lung, Rat brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: IQARVQGQRP GSPEEAAALV DGLRREPGGP RRWVTVQVQG QEVLSEKMEP SSFQPLPETE PPTPEPGPKT PPRTMQESPL GLQVKEESEV TEDSDFLESG PLAATQESVP TLLPEEAQRC GTVLDQIFPH SKTGPEGPSW REHPRALWHE EAGGIFSPGF ALQLGSISAG PGSVSPHLHV PWDLGMAGLS GQIQSPSREG GFAHALLLPS DLRSEQDPTD EDPCRGVGPA LITTRWRSPR GRSRGRPSTG G.
Binds to target promoter DNA and functions as transcription regulator. Regulates transcription from the PADI1 and CDH2 promoter. May be one regulator of transcriptional events during hemopoietic development. [UniProt].
Specifications
| MZF1 | |
| Polyclonal | |
| Unconjugated | |
| MZF1 | |
| myeloid zinc finger 1; Myeloid zinc finger protein 1; Myeloid zinc finger protein 2; myeloid-specific retinoic acid-responsive zinc finger protein; MZF; MZF1; MZF-1; MZF1B; Mzf2; Mzf-2; Zfp121; Zfp98; Zinc finger and SCAN domain-containing protein 6; zinc finger protein 121; Zinc finger protein 42; zinc finger protein 42 (myeloid-specific retinoic acid-responsive); zinc finger protein 98; Znf42; ZSCAN6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109889, 361508, 7593 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.36 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P28698 | |
| MZF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-350 of human MZF1 (NP_003413.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.