Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)-2-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH6066803
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50393-59-4
Molecular Formula C16H12N2O4S
Molecular Weight (g/mol) 328.34
MDL Number MFCD00583392
InChI Key YPAFWESEDDOFFH-UHFFFAOYSA-N
Synonym n-1-naphthyl-2-nitrobenzenesulfonamide, n-naphthalen-1-yl-2-nitrobenzenesulfonamide, naphthyl 2-nitrophenyl sulfonyl amine, 2-nitro-n-1-naphthyl benzenesulfonamide
PubChem CID 681787
IUPAC Name N-naphthalen-1-yl-2-nitrobenzenesulfonamide
SMILES [O-][N+](=O)C1=CC=CC=C1S(=O)(=O)NC1=CC=CC2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 3002217
Formula Weight 328.34
Percent Purity 97%
Chemical Name or Material N-(1-Naphthyl)-2-nitrobenzenesulfonamide
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.