Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)ethylenediamine Dihydrochloride, ≥98%, ACS Reagent, Thermo Scientific Chemicals™ PROMO

Catalog No. AC423990S24
Click to view available options
:
N-(1-Naphthyl)ethylenediamine Dihydrochloride; Packaging: Glass Bottle; Quantity: 25 g

Synonym: N1-(Naphthalen-1-yl)ethane-1,2-diamine Dihydrochloride, N-1-Naphthylethylenediamine Dihydrochloride, Bratton-Marshall Reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.17
MDL Number MFCD00012556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 98%
CAS Max % 100%
Decomposition Information 200°C
Color Beige to White or Pink
Infrared Spectrum Authentic
Assay Percent Range ≥98%
Packaging Glass Bottle
Linear Formula C10H7NHCH2CH2NH2·2HCl
Quantity 25 g
Beilstein 12, II, 699
Sensitivity (Sulfanilamide) Passes
Merck Index 15, 6491
Solubility Information Soluble in Water, Slightly Soluble in Acetone and Absolute Alcohol
Formula Weight 259.18
Percent Purity ≥98%
Grade ACS
Physical Form Powder
Chemical Name or Material N-(1-Naphthyl)ethylenediamine, dihydrochloride
Show More Show Less
Hazard Statement Causes skin irritation. May cause respiratory irritation.

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.