Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)-N-(phenylmethylene)amine, 97%, Thermo Scientific™

Catalog No. p-2705466
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC33318DA 1 g Amber Glass Bottle
AC33318EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC33318DA Supplier Thermo Scientific Chemicals Supplier No. AC33318DA
Only null left
Add to Cart

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 890-51-7
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD02235774
InChI Key NQPYSYDZNCHIQY-UHFFFAOYSA-N
Synonym n-1-naphthyl-n-phenylmethylene amine, n-benzylidenenaphthalen-1-amine, n-e-phenylmethylidene naphthalen-1-amine, 1z-1-naphthyl-2-phenyl-1-azaethene, n-naphthalen-1-yl-1-phenylmethanimine, 1z-n-naphthalen-1-yl-1-phenylmethanimine, n-benzylidene-1-napthylamine, benzylidene-a-napthylamine, maybridge3_000265, n-benzylidene-1-naphthalenamine
PubChem CID 1201463
IUPAC Name N-naphthalen-1-yl-1-phenylmethanimine
SMILES C1=CC=C(C=C1)C=NC2=CC=CC3=CC=CC=C32

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 231.3
Chemical Name or Material N-(1-naphthyl)-N-(phenylmethylene)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.