Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Chloroethyl)benzamide 99%, Thermo Scientific™

Catalog No. AC184510500
Click to view available options
:
50g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 26385-07-9
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C9H10ClNO
Linear Formula C6H5CONHCH2CH2Cl
Quantity 50g
Beilstein 09, 202
Synonym n-2-chloroethyl benzamide, benzamide, n-2-chloroethyl, acmc-1cb5c, 2-benzoylamino-ethyl chloride, fyqjuycgplfwqr-uhfffaoysa, n-2-chloroethyl benzamide 10g
InChI Key FYQJUYCGPLFWQR-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)NCCCl
IUPAC Name N-(2-chloroethyl)benzamide
Molecular Weight (g/mol) 183.635
PubChem CID 101439
Formula Weight 183.64
Percent Purity ≥98.5%
Chemical Name or Material N-(2-Chloroethyl)benzamide, 99%
Show More Show Less
EINECSNumber 247-641-4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.