Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1,3,4-thiadiazole-2-thiol, 98%

Catalog No. AC155970500
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle

CAS: 2349-67-9 | C2H3N3S2 | 133.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2349-67-9
Molecular Formula C2H3N3S2
Molecular Weight (g/mol) 133.2
MDL Number MFCD00003108
InChI Key GDGIVSREGUOIJZ-UHFFFAOYSA-N
Synonym 5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole
PubChem CID 2723847
IUPAC Name 5-amino-3H-1,3,4-thiadiazole-2-thione
SMILES C1(=NNC(=S)S1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 231°C to 236°C
Color Cream to Yellow or White
Loss on Drying 1% max. (110°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Beilstein 27, 674
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in nh4oh,soluble in dmf
Formula Weight 133.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Amino-1, 3, 4-thiadiazole-2-thiol, 98%
Show More Show Less
EINECSNumber 219-078-4
RTECSNumber XI4550000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.