Learn More
N-(2-Hydroxyethyl)ethylenediamine, 99%
Description
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.
Solubility
Fully miscible in water.
Notes
Air Sensitive & Hygroscopic. Store under inert gas. Keep away from incompatibles.

Specifications
Specifications
Melting Point | -28°C |
Density | 1.029 |
pH | 11.8 |
Boiling Point | 238°C to 240°C |
Flash Point | 125°C (257°F) |
Odor | Amine-like |
Refractive Index | 1.4861 |
Quantity | 500 g |
UN Number | UN2735 |
Beilstein | 506012 |
Show More |
Safety and Handling
Hazard Category | H314-H317-H360Df |
Hazard Statement | GHS H Statement H360-H314-H318-H317 May damage fertility or the unborn child. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P260-P264b-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
EINECSNumber | 203-867-5 |
RTECSNumber | KJ6300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.