Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)phthalimide, 99%

Catalog No. p-3758182
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 3891-07-4 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3891-07-4
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005903
InChI Key MWFLUYFYHANMCM-UHFFFAOYSA-N
Synonym 2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, n-pinacolylphthalamic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, +-n-pinacolylphthalamic acid
PubChem CID 3354762
IUPAC Name 2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
SMILES OCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 126°C to 129°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 191.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
Show More Show Less
EINECSNumber 223-434-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.