Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Aminopropyl)diethanolamine 90.0+%, TCI America™
SDP

Catalog No. A128025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A128025G 25 g
A1280500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A128025G Supplier TCI America Supplier No. A128025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4985-85-7
Molecular Formula C7H18N2O2
Molecular Weight (g/mol) 162.23
MDL Number MFCD00047971
InChI Key FKJVYOFPTRGCSP-UHFFFAOYSA-N
Synonym N,N-Bis(2-hydroxyethyl)-1,3-diaminopropane
PubChem CID 21088
IUPAC Name 2-[(3-aminopropyl)(2-hydroxyethyl)amino]ethan-1-ol
SMILES NCCCN(CCO)CCO

Specifications

Color Yellow
pH 12
Boiling Point 207°C
Quantity 25 g
UN Number 2735
Formula Weight 162.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(3-Aminopropyl)diethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.