Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-hydroxypropyl)phthalimide, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
883-44-3
C11H11NO3
205.21
MFCD00023097
BSMILTTURCQDGJ-UHFFFAOYSA-N
n-3-hydroxypropyl phthalimide,2-3-hydroxypropyl isoindoline-1,3-dione,n-3-hydroxypropyl-phthalimide,1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl,3-phthalimido-1-propanol,2-3-hydroxypropyl isoindole-1,3-dione,2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione,3-phthalimidopropanol,3-phthalimidylpropanol,acmc-209qsg
70160
2-(3-hydroxypropyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO
883-44-3
C11H11NO3
205.21
MFCD00023097
BSMILTTURCQDGJ-UHFFFAOYSA-N
n-3-hydroxypropyl phthalimide,2-3-hydroxypropyl isoindoline-1,3-dione,n-3-hydroxypropyl-phthalimide,1h-isoindole-1,3 2h-dione, 2-3-hydroxypropyl,3-phthalimido-1-propanol,2-3-hydroxypropyl isoindole-1,3-dione,2-3-hydroxypropyl-1h-isoindole-1,3 2h-dione,3-phthalimidopropanol,3-phthalimidylpropanol,acmc-209qsg
70160
2-(3-hydroxypropyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCO
Melting Point 75°C to 77°C
Color White
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Solubility Information Solubility in water: < 1g/l
Formula Weight 205.21
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material N-(3-Hydroxypropyl)phthalimide
Show More Show Less

EINECSNumber : 212-931-1