Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol

Catalog No. AAH6641422
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6641422 100 g
AAH6641414 25 g
AAH6641436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6641422 Supplier Thermo Scientific Chemicals Supplier No. H6641422
Only null left
Add to Cart

CAS: 35141-36-7 | C9H24ClNO3Si | 257.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35141-36-7
Molecular Formula C9H24ClNO3Si
Molecular Weight (g/mol) 257.83
MDL Number MFCD00054225
InChI Key FYZFRYWTMMVDLR-UHFFFAOYSA-M
Synonym n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium
PubChem CID 94462
IUPAC Name trimethyl(3-trimethoxysilylpropyl)azanium;chloride
SMILES C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]

Specifications

Color Colorless to Yellow
Density 0.930 g/mL
Flash Point 11°C (52°F)
Refractive Index 1.397
Quantity 100 g
UN Number UN1230
Beilstein 11593127
Sensitivity Hygroscopic
Formula Weight 257.83
Physical Form Liquid
Chemical Name or Material N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, in 50% methanol
Show More Show Less
Hazard Category H225-H301+H311+H331-H370
Hazard Statement GHS H Statement
H225-H311-H331-H370-H335-H302-H315-H319
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
EINECSNumber 252-393-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.