Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobenzoyl)-beta-alanine 98.0+%, TCI America™
SDP

Catalog No. A234725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A234725G 25 g
A23475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A234725G Supplier TCI America Supplier No. A234725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7377-08-4
Molecular Formula C10H11N2O3
Molecular Weight (g/mol) 207.21
MDL Number MFCD00009805
InChI Key VHAXWROFYVPXMZ-UHFFFAOYSA-M
Synonym n-4-aminobenzoyl-beta-alanine, 3-4-amino-benzoylamino-propionic acid, 3-4-aminobenzamido propanoic acid, 4-aminobenzoyl-beta-alanine, unii-ab85ljq945, 3-4-aminobenzoyl amino propanoic acid, n-4-aminophenyl carbonyl-beta-alanine, 3-4-aminophenyl formamido propanoic acid, 3-4-aminophenyl carbonylamino propanoic acid
PubChem CID 719629
IUPAC Name 3-[(4-aminophenyl)formamido]propanoate
SMILES NC1=CC=C(C=C1)C(=O)NCCC([O-])=O

Specifications

Melting Point 155°C
Color Yellow
Quantity 25 g
Formula Weight 208.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobenzoyl)-beta-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.