Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Biphenylyl)-1-naphthylamine 98.0+%, TCI America™
SDP

Catalog No. B50471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50471G 1 g
B5047200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50471G Supplier TCI America Supplier No. B50471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 446242-37-1
Molecular Formula C22H17N
Molecular Weight (g/mol) 295.385
MDL Number MFCD28138082
InChI Key KDADHQHDRSAQDY-UHFFFAOYSA-N
Synonym N-([1,1′C-Biphenyl]-4-yl)naphthalen-1-amine
PubChem CID 59029879
IUPAC Name N-(4-phenylphenyl)naphthalen-1-amine
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=CC4=CC=CC=C43

Specifications

Melting Point 149°C
Boiling Point 315°C
Quantity 1 g
Formula Weight 295.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Biphenylyl)-1-naphthylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.