Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Bromobutyl)phthalimide, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
5394-18-3
C12H12BrNO2
282.14
MFCD00005905
UXFWTIGUWHJKDD-UHFFFAOYSA-N
n-4-bromobutyl phthalimide,2-4-bromobutyl isoindoline-1,3-dione,1h-isoindole-1,3 2h-dione, 2-4-bromobutyl,2-4-bromobutyl-1h-isoindole-1,3 2h-dione,n-4-bromobutyl-phthalimide,2-4-bromobutyl isoindole-1,3-dione,2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione,n-4-bromobutyl,n-bromobutylphthalimide
93575
2-(4-bromobutyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCBr
5394-18-3
C12H12BrNO2
282.14
MFCD00005905
UXFWTIGUWHJKDD-UHFFFAOYSA-N
n-4-bromobutyl phthalimide,2-4-bromobutyl isoindoline-1,3-dione,1h-isoindole-1,3 2h-dione, 2-4-bromobutyl,2-4-bromobutyl-1h-isoindole-1,3 2h-dione,n-4-bromobutyl-phthalimide,2-4-bromobutyl isoindole-1,3-dione,2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione,n-4-bromobutyl,n-bromobutylphthalimide
93575
2-(4-bromobutyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCBr
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 76.0°C to 80.0°C
Quantity 10g
Color White to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 21, 463
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 282.14
Physical Form Powder
Percent Purity 98%
Chemical Name or Material N-(4-Bromobutyl)phthalimide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.