Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Chlorophenyl)phthalimide 98.0+%, TCI America™
SDP

Catalog No. C195825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C195825G 25 g
C19585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C195825G Supplier TCI America Supplier No. C195825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7386-21-2
Molecular Formula C14H8ClNO2
Molecular Weight (g/mol) 257.67
MDL Number MFCD00023048
InChI Key QKHKQJWODBAIMN-UHFFFAOYSA-N
Synonym n-4-chlorophenyl phthalimide, n-p-chlorophenyl phthalimide, phthalimide, n-p-chlorophenyl, 1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl, 2-4-chlorophenyl isoindoline-1,3-dione, 2-4-chlorophenyl-1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl isoindole-1,3-dione, acmc-209osn, 5-21-10-00293 beilstein handbook reference, n-4-chlorophenyl-phthalimide
PubChem CID 81867
IUPAC Name 2-(4-chlorophenyl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES ClC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 198°C
Color White
Quantity 25 g
Formula Weight 257.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Chlorophenyl)phthalimide
RTECSNumber TI4267000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.