Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Fluorobenzylidene)aniline 98.0+%, TCI America™
SDP

Catalog No. p-7129962
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 5676-81-3
Molecular Formula C13H10FN
Molecular Weight (g/mol) 199.228
MDL Number MFCD00017971
InChI Key MPRONVWLCPZXOB-UHFFFAOYSA-N
Synonym n-4-fluorobenzylidene aniline, 4-fluorobenzylideneaniline, 1-4-fluorophenyl-n-phenylmethanimine, benzenamine, n-4-fluorophenyl methylene-, e, p-fluorobenzylideneaniline, acmc-20m5v4, ambscpod_02/0603, 4-fluorophenyl methylenebenzenamine, e-n-4-fluorobenzylidene aniline, n-e-4-fluorobenzylidene aniline
PubChem CID 918392
IUPAC Name 1-(4-fluorophenyl)-N-phenylmethanimine
SMILES C1=CC=C(C=C1)N=CC2=CC=C(C=C2)F

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 176°C
Quantity 1 g
Formula Weight 199.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Fluorobenzylidene)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.