Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-(4-Fluorophenyl)glycine 98.0+%, TCI America™

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 138°C |
Color | White-Yellow |
Quantity | 5 g |
Formula Weight | 169.16 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | N-(4-Fluorophenyl)glycine |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction