Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(6-Bromohexyl)phthalimide, 97%

Catalog No. AAB2512814
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 24566-79-8 | C14H16BrNO2 | 310.191 g/mol

N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24566-79-8
Molecular Formula C14H16BrNO2
Molecular Weight (g/mol) 310.191
MDL Number MFCD00023098
InChI Key OAZFTIPKNPTDIO-UHFFFAOYSA-N
PubChem CID 141120
IUPAC Name 2-(6-bromohexyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr

Specifications

Melting Point 54°C to 60°C
Boiling Point 186°C to 188°C (0.4 mmHg)
Odor Odorless
Quantity 25 g
Beilstein 186875
Synonym n-6-bromohexyl phthalimide, 2-6-bromohexyl isoindoline-1,3-dione, n-6-bromohexyl-phthalimide, 2-6-bromohexyl-1h-isoindole-1,3 2h-dione, 2-6-bromohexyl isoindole-1,3-dione, 2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-6-bromo-hexyl-isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-6-bromohexyl, 1h-isoindole-1,3 2h-dione, 2-6-bromohexyl, 6-n-phthalimido hexyl bromide
Solubility Information Insoluble in water.
Formula Weight 310.19
Percent Purity 97%
Chemical Name or Material N-(6-Bromohexyl)phthalimide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.