Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. F10311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10311G 1 g
F10315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10311G Supplier TCI America Supplier No. F10311G

Chemical Identifiers

CAS 204376-55-6
Molecular Formula C20H19NO3
Molecular Weight (g/mol) 321.38
MDL Number MFCD00673794
InChI Key AMWDMRLGMSQZTK-UHFFFAOYSA-N
Synonym N-Fmoc-4-piperidone
PubChem CID 736303
IUPAC Name (9H-fluoren-9-yl)methyl 4-oxopiperidine-1-carboxylate
SMILES O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)N1CCC(=O)CC1

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 321.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.