Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-beta-alanine, 97%

Catalog No. AAH5020803
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH5020803

Supplier: Thermo Scientific Chemicals H5020803

Only null left
Add to Cart

CAS: 3025-95-4 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3025-95-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00037296
InChI Key LJLLAWRMBZNPMO-UHFFFAOYSA-N
Synonym n-acetyl-beta-alanine, n-acetyl-b-alanine, ac-beta-ala-oh, n-acetyl-, a-alanine, 3-acetylamino propanoic acid, n-acetyl-?-alanine, acetyl-beta-alanine, n-acetyl-ss-alanine, ac--ala-oh, beta-alanine, n-acetyl
PubChem CID 76406
ChEBI CHEBI:16682
IUPAC Name 3-acetamidopropanoic acid
SMILES CC(=O)NCCC(O)=O

Specifications

Melting Point 77°C
Linear Formula CH3CONHCH2CH2CO2H
Quantity 1 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material N-Acetyl-beta-alanine
EINECSNumber 221-185-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.