Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-glucosamine, 96.0-102.0%, Spectrum™ Chemical
SDP

Catalog No. 18611892 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-892 100 g Poly Bottle
18-611-890 25 g Poly Bottle
18-611-894 500 g Poly Bottle
18-611-891 5 kg Poly Pail
18-611-893 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-892 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1269100GM
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
pH 6.0 to 8.0
Packaging Poly Bottle
Quantity 100 g
Specific Rotation +39.0° to +43.0°
Percent Purity 96 to 102.0%
Grade Reagent

CAUTION: For manufacturing, laboratory or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.