Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Acetyl-D-Glucosamine Kinase/NAGK Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142694AF405
Description
N-Acetyl-D-Glucosamine Kinase/NAGK Polyclonal antibody specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human samples. It is validated for Western BlotSpecifications
N-Acetyl-D-Glucosamine Kinase/NAGK | |
Polyclonal | |
Western Blot | |
EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910 | |
The immunogen for this product maps to a region between residue 50 and 100 of human N-acetylglucosamine kinase using the numbering given in entry NP_060037.2 (GeneID 55577). | |
0.1 mL | |
metabolism | |
55577 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction