Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99%
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.
Solubility
Soluble in water, ethanol, ethyl acetate.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.
Solubility
Soluble in water, ethanol, ethyl acetate.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 113°C to 117°C |
Beilstein | 1725554 |
Merck Index | 14,96 |
Solubility Information | Soluble in water,ethanol,ethyl acetate. |
IUPAC Name | 2-acetamido-4-methylsulfanylbutanoic acid |
Quantity | 1000 g |
Formula Weight | 191.25 |
Percent Purity | 99% |
Chemical Name or Material | N-Acetyl-DL-methionine |
Safety and Handling
EINECSNumber | 214-224-3 |
RTECSNumber | PD0500000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction