Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodomethane, 99%, stabilized
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -66.0°C |
Density | 2.2800g/mL |
Boiling Point | 42.5°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Refractive Index | 1.5290 to 1.5310 |
Linear Formula | CH3I |
Quantity | 500 g |
Specific Gravity | 2.28 |
Solubility Information | Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | GHS Signal Word: Danger |
missing translation for 'healthHazard2' | GHS H Statement Toxic if swallowed. Harmful in contact with skin. Causes skin irritation. Toxic if inhaled. May cause respiratory irritation. Suspected of causing cancer. |
missing translation for 'healthHazard3' | GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of s |
EINECSNumber | 200-819-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction