Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N-Acetyl-L-histidine monohydrate

Catalog No. AAJ6565706
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 39145-52-3 | C8H13N3O4 | 215.209 g/mol

N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for NÃŽ±-acetylhistidine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for Nα-acetylhistidine synthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39145-52-3
Molecular Formula C8H13N3O4
Molecular Weight (g/mol) 215.209
MDL Number MFCD00149320
InChI Key PSWSDQRXCOJSFC-FJXQXJEOSA-N
Synonym ac-his-oh.h2o, n-acetyl-l-histidine hydrate, n-acetyl-l-histidine monohydrate, unii-pq2tc3x11o, s-2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, pq2tc3x11o, acetyl histidine, 2s-2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, l-n-acetylhistidine monohydrate
PubChem CID 2724380
IUPAC Name (2S)-2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
SMILES CC(=O)NC(CC1=CN=CN1)C(=O)O.O

Specifications

Melting Point 163°C
Color White
Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 215.21 (197.09 Anhydrous)
Optical Rotation +47° (c=1 in Water)
Physical Form Powder
Chemical Name or Material N-Acetyl-L-histidine monohydrate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.