Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-ornithine, 95%

Catalog No. AAH6664803
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 6205-08-9 | C7H14N2O3 | 174.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6205-08-9
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.20
MDL Number MFCD00065115
InChI Key JRLGPAXAGHMNOL-UHFFFAOYNA-N
PubChem CID 439232
ChEBI CHEBI:16543
IUPAC Name (2S)-2-acetamido-5-aminopentanoic acid
SMILES CC(=O)NC(CCCN)C(=O)O

Specifications

Synonym n-acetylornithine, ac-orn-oh, n2-acetyl-l-ornithine, n 2-acetyl-l-ornithine, acetylornithine, 2s-2-acetamido-5-aminopentanoic acid, n∼2∼-acetyl-l-ornithine, aor, ornithine, n2-acetyl, nalpha-acetyl-l-ornithine
Quantity 1 g
Formula Weight 174.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Acetyl-L-ornithine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.