Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-tyrosine ethyl ester monohydrate, 97%

Catalog No. aaa1157706
Click to view available options
Quantity:
5 g
25 g

CAS: 36546-50-6 | C13H19NO5 | 269.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36546-50-6
Molecular Formula C13H19NO5
Molecular Weight (g/mol) 269.30
MDL Number MFCD00149093
InChI Key YWAVLHZJMWEYTA-HVPILOLGNA-N
Synonym n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
PubChem CID 2723594
IUPAC Name ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
SMILES O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O

Specifications

Melting Point 80°C to 81°C
Quantity 5 g
Formula Weight 269.30 (251.28 Anhydrous)
Percent Purity 97%
Sensitivity Moisture sensitive
Chemical Name or Material N-Acetyl-L-tyrosine ethyl ester monohydrate
EINECSNumber 212-663-5
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.