Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-O-tert-butyl-L-serine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6523906
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77285-09-7
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00236770
InChI Key COMGVZVKADFEPL-UHFFFAOYNA-N
Synonym ac-ser tbu-oh, l-serine,n-acetyl-o-1,1-dimethylethyl, ambotzaaa1904, n-acetyl-o-t-butyl-l-serine, 2s-3-tert-butoxy-2-acetamidopropanoic acid
PubChem CID 7019612
IUPAC Name 3-(tert-butoxy)-2-acetamidopropanoic acid
SMILES CC(=O)NC(COC(C)(C)C)C(O)=O

Specifications

Color White
Beilstein 9047283
Quantity 5 g
Formula Weight 203.24
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-O-tert-butyl-L-serine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.