Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ N-Acetylcysteine
GREENER_CHOICESDP

Catalog No. 7874100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-741-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 78-741-00 Supplier Tocris Bioscience™ Supplier No. 7874/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glutathione (GSH) precursor; maintains stem cell function in culture.

N-Acetylcysteine is a cell-permeable antioxidant and a precursor of reduced glutathione (GSH) (Cat. No. 5219), with anti-inflammatory, mucolytic, and antiviral activities. N-Acetylcysteine can be used in organoid culture media for non-small cell lung and colorectal cancer organoids, expansion media for mouse/human liver and pancreas 3D organoids and LWRN media for colonic 3D organoids from human biopsies. It is a component of S7 medium in the optimized protocol for generation of functional stem cell-derived islets (SC-islets). N-Acetylcysteine can be orally administered.

Specifications

Chemical Name or Material (2R)-2-Acetamido-3-sulfanylpropanoic acid
CAS 616-91-1
Quantity 100 mg
Target Additional Antioxidant Compounds
Molecular Formula C5H9NO3S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.