Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ N-Acetylcysteine amide
GREENER_CHOICESDP

Catalog No. 561950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
561950 50 mg
561910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 561950 Supplier Tocris Bioscience™ Supplier No. 5619/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Glutathione (GSH) precursor; neuroprotective

N-Acetylcysteine amide is a glutathione (GSH) precursor and cell-permeable antioxidant. N-Acetylcysteine amide replenishes intracellular Glutathione (GSH). N-Acetylcysteine amide directly reduces intracellular Glutathione disulfide (GSSG) to GSH without glutathione peroxidase. N-Acetylcysteine amide has anti-inflammatory activity through regulation of activation of NF-κB and HIF-1α, as well as modulation of reactive oxygen species. N-Acetylcysteine amide improves neuronal mitochondrial bioenergetics, reduces tissue damage and enhances functional recovery following spinal cord injury in rats. N-Acetylcysteine amide also enhances behavioral recovery in rats following traumatic brain injury. Neuroprotective.

For more information about how N-Acetylcysteine amide may be used, see our protocol: 3D Culture of Lung Alveolar Cells

Specifications

Chemical Name or Material (2R)-2-(Acetylamino)-3-mercaptopropanamide
CAS 38520-57-9
Quantity 50 mg
Target Additional Antioxidant Compounds
Molecular Formula C5H10N2O2S
Formula Weight 162.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.