Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Acetylmannosamine Kinase/GNE Antibody (SR2196), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32258750UL
Description
N-Acetylmannosamine Kinase/GNE Monoclonal antibody specifically detects N-Acetylmannosamine Kinase/GNE in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
N-Acetylmannosamine Kinase/GNE | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
SR2196 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, DMRV, GLCNENM, glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase, IBM2, N-acylmannosamine kinase, Uae1, UDP-GlcNAc-2-epimerase/ManAc kinase, UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase | |
A synthesized peptide derived from human N-Acetylmannosamine Kinase/GNE (Uniprot #: Q9Y223) | |
50 μg | |
Cardiovascular Biology, Cell Biology, Signal Transduction | |
10020 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction