Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetylmorpholine, 98%

Click to view available options
Quantity:
25g
100g
1696-20-4
C6H11NO2
129.159
MFCD00006171
KYWXRBNOYGGPIZ-UHFFFAOYSA-N
4-acetylmorpholine,n-acetylmorpholine,1-morpholinoethanone,morpholine, 4-acetyl,n-acetylmorfolin,n-acetylmorfolin czech,ethanone, 1-4-morpholinyl,n-acetyl morpholine,1-morpholin-4-yl ethan-1-one,acetylmorpholine
15543
1-morpholin-4-ylethanone
CC(=O)N1CCOCC1
1696-20-4
C6H11NO2
129.159
MFCD00006171
KYWXRBNOYGGPIZ-UHFFFAOYSA-N
4-acetylmorpholine,n-acetylmorpholine,1-morpholinoethanone,morpholine, 4-acetyl,n-acetylmorfolin,n-acetylmorfolin czech,ethanone, 1-4-morpholinyl,n-acetyl morpholine,1-morpholin-4-yl ethan-1-one,acetylmorpholine
15543
1-morpholin-4-ylethanone
CC(=O)N1CCOCC1
Flash Point >110°C (230°F)
Quantity 100g
Boiling Point 245°C to 246°C
Melting Point 9°C to 10°C
Beilstein 111971
Formula Weight 129.16
Percent Purity 98%
Chemical Name or Material N-Acetylmorpholine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QD7000000

TSCA : TSCA