Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-acetylprocainamide Monoclonal Antibody (NAPA11-128.2.1), QED Bioscience™
SDP

Catalog No. 502277781
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7781 500 μg
50-227-7780 100 μg
50-227-7779 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7781 Supplier Leinco Technologies Inc Supplier No. 16908500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

N-acetylprocainamide Monoclonal antibody specifically detects N-acetylprocainamide in Chemical samples. It is validated for ELISA

Specifications

Antigen N-acetylprocainamide
Applications ELISA
Classification Monoclonal
Clone NAPA11-128.2.1
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Acecainida; Acecainide; Acecainidum; Acekainid; C15H23N3O2; N-Acetyloprokainamid
Host Species Mouse
Immunogen N-Acetylprocainamide-KLH.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.