Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylsulfanilyl chloride, 99%

Catalog No. p-50182
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle

CAS: 121-60-8 | C8H8ClNO3S | 233.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-60-8
Molecular Formula C8H8ClNO3S
Molecular Weight (g/mol) 233.67
MDL Number MFCD00007442
InChI Key GRDXCFKBQWDAJH-UHFFFAOYSA-N
Synonym n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride
PubChem CID 8481
IUPAC Name 4-acetamidobenzenesulfonyl chloride
SMILES CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 145.0°C to 148.0°C
Color Beige-Cream to White
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula 4-(CH3CONH)C6H4SO2Cl
Quantity 100 g
Beilstein 14, 702
Fieser 01,3
Merck Index 15, 95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethylene dichloride,,alcohol,ether
Formula Weight 233.67
Percent Purity 99%
Physical Form Granular Crystalline Powder or Crystals
Chemical Name or Material N-Acetylsulfanilyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 204-485-1
RTECSNumber DB8837500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.