Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Allylaniline, 95%

Catalog No. AAH5582122
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 589-09-3 | C9H11N | 133.19 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep away from oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-09-3
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008638
InChI Key LQFLWKPCQITJIH-UHFFFAOYSA-N
Synonym n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
PubChem CID 68525
SMILES C=CCNC1=CC=CC=C1

Specifications

Color Colorless to Brown
Density 0.982 g/mL
Boiling Point 218°C to 220°C
Flash Point 89°C (192°F)
Refractive Index 1.563
Linear Formula C6H5NHCH2CH=CH2
Quantity 100 g
Solubility Information Insoluble in water.
IUPAC Name N-(prop-2-en-1-yl)aniline
Formula Weight 133.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Allylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H227-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 209-633-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.