Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Allylaniline 99.0+%, TCI America™
SDP

Catalog No. A0880100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0880100ML 100 mL
A088025ML 25 mL
A0880500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0880100ML Supplier TCI America Supplier No. A0880100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 589-09-3
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008638
InChI Key LQFLWKPCQITJIH-UHFFFAOYSA-N
Synonym n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
PubChem CID 68525
IUPAC Name N-(prop-2-en-1-yl)aniline
SMILES C=CCNC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 100 mL
Formula Weight 133.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Allylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.